missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylphenol, 97%, Thermo Scientific Chemicals

£26.10 - £220.00

Chemical Identifiers

CAS 2362-12-1
MDL Number MFCD00055435
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15485017
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32307.03
1 g
£26.10
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15495017
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32307.06
5 g
£69.60
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15405027
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32307.14
25 g
£220.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Bromo-2-methylphenol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methylphenol, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

2362-12-1
MFCD00055435

Specifications

2362-12-1
Phenol-like
MFCD00055435
Slightly soluble in water.
CC1=C(C=CC(=C1)Br)O
187.04
187.04
4-Bromo-2-methylphenol
62°C to 66°C
C7H7BrO
4-bromo-o-cresol, 2-methyl-4-bromophenol, 4-bromo-2-methyl-phenol, o-cresol, 4-bromo, phenol, 4-bromo-2-methyl, 2-hydroxy-5-bromotoluene, p-bromocresol, 4-bromo-cresol, pubchem7106, acmc-1chy8
IWJGMJHAIUBWKT-UHFFFAOYSA-N
4-bromo-2-methylphenol
16896
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

missing translation for 'rtecsNumber' : GO6868000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only