missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylpyridine, 97%, Thermo Scientific Chemicals

Product Code. 15463807
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Bromo-2-methylpyridine is used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reductive coupling, side chain oxidation and esterification. Further, it serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals and in dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methylpyridine is used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reductive coupling, side chain oxidation and esterification. Further, it serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals and in dyes.

Solubility
Miscible with dichloromethane. Immiscible with water.

Notes
Light and moisture sensitive. Incompatible with strong oxidizing agents, reducing agents, strong acids and bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22282-99-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD03788196
InChI Key JFBMFWHEXBLFCR-UHFFFAOYSA-N
Synonym 4-bromo-2-picoline, 2-methyl-4-bromopyridine, 4-bromo-2-methyl-pyridine, pyridine, 4-bromo-2-methyl, 4-bromopicoline, zlchem 422, pubchem4035, acmc-209fui, 4-bromo-methyl-pyridine
PubChem CID 2762835
IUPAC Name 4-bromo-2-methylpyridine
SMILES CC1=CC(Br)=CC=N1

Specifications

Density 1.45
Boiling Point 76°C (14 mmHg)
Flash Point 79°C (174°F)
Odor Characteristic
Refractive Index 1.556
Quantity 250 mg
Solubility Information Miscible with dichloromethane. Immiscible with water.
Formula Weight 172.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylpyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'