missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitroaniline, 98%, Thermo Scientific Chemicals

Product Code. 11431050
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875-51-4
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.02
MDL Number MFCD00041312
InChI Key ZCWBZRBJSPWUPG-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-2-nitro, 2-nitro-4-bromoaniline, 4-bromo-o-nitroaniline, p-bromo-o-nitroaniline, 4-bromo-2-nitro-phenylamine, 4-bromo-2-nitrobenzeneamine, aniline, 4-bromo-2-nitro, 4-bromo-2-nitrobenzenamine, 1-amino-4-bromo-2-nitrobenzene, 4-bromo-2-nitro-aniline
PubChem CID 70132
IUPAC Name 4-bromo-2-nitroaniline
SMILES NC1=CC=C(Br)C=C1[N+]([O-])=O

Specifications

Melting Point 111°C to 114°C
Quantity 100 g
Formula Weight 217.02
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-nitroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'