missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethoxy)benzene-1-thiol, ≥95%, Thermo Scientific™

Catalog Number 10591123
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
175278-15-6
C7H4BrF3OS
273.07
MFCD00203474
WWZNHDMEBZHFTK-UHFFFAOYSA-N
4-bromo-2-trifluoromethoxy thiophenol, 4-bromo-2-trifluoromethoxy benzenethiol, 4-bromo-2-trifluoromethoxy benzene-1-thiol, benzenethiol,4-bromo-2-trifluoromethoxy, pubchem2840, 4-bromo-2-trifluoromethoxy-benzenethiol, 4-bromanyl-2-trifluoromethyloxy benzenethiol
2736425
4-bromo-2-(trifluoromethoxy)benzenethiol
FC(F)(F)OC1=C(S)C=CC(Br)=C1
This item is not returnable. View return policy
175278-15-6
C7H4BrF3OS
273.07
MFCD00203474
WWZNHDMEBZHFTK-UHFFFAOYSA-N
4-bromo-2-trifluoromethoxy thiophenol, 4-bromo-2-trifluoromethoxy benzenethiol, 4-bromo-2-trifluoromethoxy benzene-1-thiol, benzenethiol,4-bromo-2-trifluoromethoxy, pubchem2840, 4-bromo-2-trifluoromethoxy-benzenethiol, 4-bromanyl-2-trifluoromethyloxy benzenethiol
2736425
4-bromo-2-(trifluoromethoxy)benzenethiol
FC(F)(F)OC1=C(S)C=CC(Br)=C1
CAS Min % 95.0
Assay Percent Range 95%
Quantity 1 g
Formula Weight 273.07
Percent Purity 95%
Chemical Name or Material 4-Bromo-2-(trifluoromethoxy)benzene-1-thiol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'