missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dihydroxybenzoic acid, 97+%, Thermo Scientific Chemicals

Product Code. 11493158
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16534-12-6
Molecular Formula C7H5BrO4
Molecular Weight (g/mol) 233.017
MDL Number MFCD00002513
InChI Key NUTRHYYFCDEALP-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-3,5-dihydroxy, 4-bromo-3,5-resorcylic acid, 4-bromo-alpha-resorcylic acid, 4-bromo-3,5-dihydroxybenzic acid, .alpha.-resorcylic acid, 4-bromo, 4-bromo-3,5-dihydroxy-benzoic acid, alpha-resorcylic acid, 4-bromo, pubchem8196, acmc-209dsh, ksc177s3r
PubChem CID 86023
IUPAC Name 4-bromo-3,5-dihydroxybenzoic acid
SMILES C1=C(C=C(C(=C1O)Br)O)C(=O)O

Specifications

Melting Point 268°C to 272°C
Quantity 500 g
Beilstein 2368265
Sensitivity Light sensitive
Formula Weight 233.02
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-3,5-dihydroxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'