Learn More
4-Bromo-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
£51.10 - £1105.00
Chemical Identifiers
CAS | 7463-51-6 |
---|---|
Molecular Formula | C8H9BrO |
Molecular Weight (g/mol) | 201.06 |
MDL Number | MFCD00002315 |
InChI Key | WMUWDPLTTLJNPE-UHFFFAOYSA-N |
Synonym | 4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606 |
PubChem CID | 81970 |
IUPAC Name | 4-bromo-3,5-dimethylphenol |
SMILES | CC1=CC(=CC(=C1Br)C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10449780
|
Thermo Scientific Acros
106870100 |
10 g |
£51.10
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10459780
|
Thermo Scientific Acros
106870250 |
25 g |
£102.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10269513
|
Thermo Scientific Acros
106871000 |
100 g |
£293.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11934681
|
Thermo Scientific Acros
106875000 |
500 g |
£1105.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7463-51-6 | |
201.06 | |
WMUWDPLTTLJNPE-UHFFFAOYSA-N | |
81970 | |
CC1=CC(=CC(=C1Br)C)O |
C8H9BrO | |
MFCD00002315 | |
4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606 | |
4-bromo-3,5-dimethylphenol |
Specifications
7463-51-6 | |
100.0 | |
Colorless to Pink | |
99% | |
C8H9BrO | |
MFCD00002315 | |
4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606 | |
CC1=CC(=CC(=C1Br)C)O | |
201.06 | |
201.06 | |
Crystals or Powder |
98.5 | |
113°C to 116°C | |
Authentic | |
Glass bottle | |
BrC6H2(CH3)2OH | |
06,I,244 | |
WMUWDPLTTLJNPE-UHFFFAOYSA-N | |
4-bromo-3,5-dimethylphenol | |
81970 | |
99% | |
4-Bromo-3, 5-dimethylphenol, 99% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-255-8
- 4-Bromo-3,5-dimethylphenol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only