missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylanisole, 97%, Thermo Scientific Chemicals

£36.38 - £468.00

Chemical Identifiers

CAS 27060-75-9
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00060687
InChI Key BLZNSXFQRKVFRP-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxytoluene, 4-bromo-3-methylanisole, benzene, 1-bromo-4-methoxy-2-methyl, 4-bromo-3-methyl-anisole, unii-okr329132j, 1-bromo-2-methyl-4-methoxybenzene, 1-brom-4-methoxy-2-methylbenzol, pubchem3796, 3-methyl-4-bromoanisole, acmc-1cg4z
PubChem CID 117915
IUPAC Name 1-bromo-4-methoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)OC)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11435073
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12318.09
10 g
£36.38
10g
Please to purchase this item. Need a web account? Register with us today!
11445073
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12318.18
50 g
£115.58
50g
Please to purchase this item. Need a web account? Register with us today!
11459423
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12318.30
250 g
£468.00
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

27060-75-9
201.063
BLZNSXFQRKVFRP-UHFFFAOYSA-N
117915
CC1=C(C=CC(=C1)OC)Br
C8H9BrO
MFCD00060687
2-bromo-5-methoxytoluene, 4-bromo-3-methylanisole, benzene, 1-bromo-4-methoxy-2-methyl, 4-bromo-3-methyl-anisole, unii-okr329132j, 1-bromo-2-methyl-4-methoxybenzene, 1-brom-4-methoxy-2-methylbenzol, pubchem3796, 3-methyl-4-bromoanisole, acmc-1cg4z
1-bromo-4-methoxy-2-methylbenzene

Specifications

27060-75-9
111°C to 112°C (15 mmHg)
C8H9BrO
MFCD00060687
2-bromo-5-methoxytoluene, 4-bromo-3-methylanisole, benzene, 1-bromo-4-methoxy-2-methyl, 4-bromo-3-methyl-anisole, unii-okr329132j, 1-bromo-2-methyl-4-methoxybenzene, 1-brom-4-methoxy-2-methylbenzol, pubchem3796, 3-methyl-4-bromoanisole, acmc-1cg4z
CC1=C(C=CC(=C1)OC)Br
201.063
201.06
4-Bromo-3-methylanisole
1.45
>110°C (230°F)
1.561
1933865
BLZNSXFQRKVFRP-UHFFFAOYSA-N
1-bromo-4-methoxy-2-methylbenzene
117915
97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only