missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitrobenzoic acid, 95%, Thermo Scientific Chemicals

Product Code. 15478568
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6319-40-0
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00272137
InChI Key RVCTZJVBWNFYRU-UHFFFAOYSA-N
Synonym 4-bromo-3-nitrobenzoic acid, 3-nitro-4-bromobenzoic acid, benzoic acid, 4-bromo-3-nitro, 4-bromo-3-nitro-benzoic acid, zlchem 531, pubchem2194, acmc-209ndq, ksc353s1b, 4-bromo-3-nitrobenzoic acid #
PubChem CID 232986
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Br

Specifications

Melting Point 199°C to 206°C
Color White to Yellow
Quantity 5 g
Formula Weight 246.01
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Bromo-3-nitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.