Learn More
4-Bromo-5-fluoro-2-nitrotoluene, 98%, Thermo Scientific™
£23.00 - £58.80
Chemical Identifiers
CAS | 224185-19-7 |
---|---|
Molecular Formula | C7H5BrFNO2 |
Molecular Weight (g/mol) | 234.02 |
MDL Number | MFCD04039299 |
InChI Key | MTTNTJRJOFVWSY-UHFFFAOYSA-N |
Synonym | 4-bromo-5-fluoro-2-nitrotoluene, 4-bromo-3-fluoro-6-nitrotoluene, 1-bromo-2-fluoro-4-methyl-5-nitro-benzene, benzene, 1-bromo-2-fluoro-4-methyl-5-nitro, 2-nitro-4-bromo-5-fluorotoluene, 4-bromo-5-fluoro-1-methyl-2-nitrobenzene, pubchem3991, zlchem 1176, acmc-1ceig, ksc549i4p |
PubChem CID | 2779365 |
IUPAC Name | 1-bromo-2-fluoro-4-methyl-5-nitrobenzene |
SMILES | CC1=CC(F)=C(Br)C=C1[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11441850
|
Thermo Scientific Alfa Aesar
H27552.03 |
1 g |
£23.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11451850
|
Thermo Scientific Alfa Aesar
H27552.06 |
5 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
224185-19-7 | |
234.02 | |
MTTNTJRJOFVWSY-UHFFFAOYSA-N | |
2779365 | |
CC1=CC(F)=C(Br)C=C1[N+]([O-])=O |
C7H5BrFNO2 | |
MFCD04039299 | |
4-bromo-5-fluoro-2-nitrotoluene, 4-bromo-3-fluoro-6-nitrotoluene, 1-bromo-2-fluoro-4-methyl-5-nitro-benzene, benzene, 1-bromo-2-fluoro-4-methyl-5-nitro, 2-nitro-4-bromo-5-fluorotoluene, 4-bromo-5-fluoro-1-methyl-2-nitrobenzene, pubchem3991, zlchem 1176, acmc-1ceig, ksc549i4p | |
1-bromo-2-fluoro-4-methyl-5-nitrobenzene |
Specifications
224185-19-7 | |
C7H5BrFNO2 | |
4-bromo-5-fluoro-2-nitrotoluene, 4-bromo-3-fluoro-6-nitrotoluene, 1-bromo-2-fluoro-4-methyl-5-nitro-benzene, benzene, 1-bromo-2-fluoro-4-methyl-5-nitro, 2-nitro-4-bromo-5-fluorotoluene, 4-bromo-5-fluoro-1-methyl-2-nitrobenzene, pubchem3991, zlchem 1176, acmc-1ceig, ksc549i4p | |
CC1=CC(F)=C(Br)C=C1[N+]([O-])=O | |
234.02 | |
234.02 | |
4-Bromo-5-fluoro-2-nitrotoluene |
59°C to 63°C | |
MFCD04039299 | |
MTTNTJRJOFVWSY-UHFFFAOYSA-N | |
1-bromo-2-fluoro-4-methyl-5-nitrobenzene | |
2779365 | |
98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures