missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-DL-phenylalanine, 98+%, Thermo Scientific Chemicals

Product Code. 11327865
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14091-15-7
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.09
MDL Number MFCD00002599
InChI Key PEMUHKUIQHFMTH-UHFFFAOYSA-N
Synonym 2-amino-3-4-bromophenyl propanoic acid, p-bromo-dl-phenylalanine, 4-bromo-dl-phenylalanine, dl-4-br-phe-oh, 4-bromophenylalanine, 4-bromo-phenylalanine, h-dl-phe 4-br-oh, 2-amino-3-4-bromophenyl propionic acid, 2-amino-3-4-bromo-phenyl-propionic acid, p-br-phenylalanine
PubChem CID 85681
IUPAC Name 2-amino-3-(4-bromophenyl)propanoic acid
SMILES NC(CC1=CC=C(Br)C=C1)C(O)=O

Specifications

Melting Point ∽262°C (decomposition)
Beilstein 2212158
Quantity 250 mg
Odor Odorless
Formula Weight 244.09
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-DL-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'