missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-methylaniline, 97%, Thermo Scientific Chemicals

Product Code. 15421478
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6911-87-1
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD03030391
InChI Key AYVPVDWQZAAZCM-UHFFFAOYSA-N
PubChem CID 2757052
IUPAC Name 4-bromo-N-methylaniline
SMILES CNC1=CC=C(Br)C=C1

Specifications

Density 1.482 g/mL
Boiling Point 259°C to 260°C
Refractive Index 1.613
Quantity 25 g
Synonym 4-bromo-n-methylbenzenamine, benzenamine, 4-bromo-n-methyl, n-methyl-4-bromoaniline, benzenamine,4-bromo-n-methyl, 4bromo-n-methylaniline, 4-bromo-n-methyl-aniline, n-methyl-4-bromo-aniline, acmc-1cfc7, 4-bromanyl-n-methyl-aniline, 4-bromo-phenyl-methyl-amine
Formula Weight 186.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Bromo-N-methylaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'