Learn More
4'-Bromoacetanilide, 98%, Thermo Scientific Chemicals
£39.80 - £137.00
Chemical Identifiers
CAS | 103-88-8 |
---|---|
Molecular Formula | C8H8BrNO |
Molecular Weight (g/mol) | 214.06 |
MDL Number | MFCD00000092 |
InChI Key | MSLICLMCQYQNPK-UHFFFAOYSA-N |
Synonym | 4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide |
PubChem CID | 7683 |
IUPAC Name | N-(4-bromophenyl)acetamide |
SMILES | CC(=O)NC1=CC=C(Br)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387895
|
Thermo Scientific Alfa Aesar
L02924.18 |
50 g |
£39.80
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397895
|
Thermo Scientific Alfa Aesar
L02924.30 |
250 g |
£137.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4'-Bromoacetanilide is used as an internal standard for phenylurea and the respective metabolic products in oysters. It has also been used as a reagent in the production of a new ligand to form a coordination compex between gadolinium(III) and T=titanium(IV) oxide. It was used in the synthesis of new ligand for anchoring Gd(III) chelates onto TiO2 surface.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Bromoacetanilide is used as an internal standard for phenylurea and the respective metabolic products in oysters. It has also been used as a reagent in the production of a new ligand to form a coordination compex between gadolinium(III) and T=titanium(IV) oxide. It was used in the synthesis of new ligand for anchoring Gd(III) chelates onto TiO2 surface.
Solubility
Moderately soluble in alcohol. Insoluble in water.
Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
Chemical Identifiers
103-88-8 | |
214.06 | |
MSLICLMCQYQNPK-UHFFFAOYSA-N | |
7683 | |
CC(=O)NC1=CC=C(Br)C=C1 |
C8H8BrNO | |
MFCD00000092 | |
4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide | |
N-(4-bromophenyl)acetamide |
Specifications
103-88-8 | |
Odorless | |
MFCD00000092 | |
14,1399 | |
Moderately soluble in alcohol. Insoluble in water. | |
CC(=O)NC1=CC=C(Br)C=C1 | |
214.06 | |
214.06 | |
4'-Bromoacetanilide |
166°C to 171°C | |
C8H8BrNO | |
2208091 | |
4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide | |
MSLICLMCQYQNPK-UHFFFAOYSA-N | |
N-(4-bromophenyl)acetamide | |
7683 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 203-154-9
missing translation for 'rtecsNumber' : AD9625000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only