missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromoacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 10509810
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-90-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000105
InChI Key WYECURVXVYPVAT-UHFFFAOYSA-N
Synonym 4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo
PubChem CID 7466
IUPAC Name 1-(4-bromophenyl)ethanone
SMILES CC(=O)C1=CC=C(Br)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48°C to 52°C
Color White to Yellow
Boiling Point 255°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4COCH3
Quantity 25 g
Beilstein 07,283
Merck Index 15,141
Solubility Information Solubility in water: insoluble
Formula Weight 199.05
Percent Purity 98%
Physical Form Crystalline Flakes or Crystals
Chemical Name or Material 4'-Bromoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 4''-Bromoacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'