missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoaniline, 98+%, Thermo Scientific Chemicals

Product Code. 11472088
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.

Solubility
Soluble in alcohol and ether. Insoluble in cold water.

Notes
Store in a cool place. Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-40-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.025
MDL Number MFCD00007822
InChI Key WDFQBORIUYODSI-UHFFFAOYSA-N
Synonym p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
PubChem CID 7807
IUPAC Name 4-bromoaniline
SMILES C1=CC(=CC=C1N)Br

Specifications

Melting Point 60°C to 65°C
Boiling Point 222°C to 224°C
Assay Percent Range ≥98%
Odor Characteristic
Quantity 1000 g
UN Number UN2811
Beilstein 742031
Merck Index 14,1404
Solubility Information Soluble in alcohol and ether. Insoluble in cold water.
Formula Weight 172.03
Percent Purity ≥98%
Chemical Name or Material 4-Bromoaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'