missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoanisole, 98%, Thermo Scientific Chemicals

£25.73 - £77.18

Chemical Identifiers

CAS 104-92-7
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00000097
InChI Key QJPJQTDYNZXKQF-UHFFFAOYSA-N
Synonym 4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
PubChem CID 7730
ChEBI CHEBI:47257
IUPAC Name 1-bromo-4-methoxybenzene
SMILES COC1=CC=C(C=C1)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10254130
View Documents Promotion Details
Thermo Scientific Acros
106631000
100 mL
£25.73
100mL
Please to purchase this item. Need a web account? Register with us today!
15720789
View Documents
Thermo Scientific Acros
106635000
500 mL
£77.18
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

104-92-7
187.04
QJPJQTDYNZXKQF-UHFFFAOYSA-N
7730
1-bromo-4-methoxybenzene
C7H7BrO
MFCD00000097
4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
CHEBI:47257
COC1=CC=C(C=C1)Br

Specifications

104-92-7
100.0
Colorless to Yellow
223.0°C
Authentic
Glass bottle
BrC6H4OCH3
MFCD00000097
10, 1400
4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
QJPJQTDYNZXKQF-UHFFFAOYSA-N
1-bromo-4-methoxybenzene
7730
187.04
Liquid
97.5
9.0°C to 10.0°C
1.4900g/mL
94°C
97.5% min. (GC)
C7H7BrO
1.5630 to 1.5650
06, 199
1.49
Solubility in water: immiscible
COC1=CC=C(C=C1)Br
187.04
CHEBI:47257
98%
4-Bromoanisole, 98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 203-252-1

missing translation for 'rtecsNumber' : BZ8501000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only