missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-bromobenzaldehyde, Thermo Scientific™
£9.20 - £12.70
Chemical Identifiers
CAS | 1122-91-4 |
---|---|
Molecular Formula | C7H5BrO |
Molecular Weight (g/mol) | 185.02 |
InChI Key | ZRYZBQLXDKPBDU-UHFFFAOYSA-N |
Synonym | p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd |
PubChem CID | 70741 |
IUPAC Name | 4-bromobenzaldehyde |
SMILES | C1=CC(=CC=C1C=O)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10075602
|
Thermo Scientific Maybridge
SB01132EA |
10 g |
£9.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10080364
|
Thermo Scientific Maybridge
SB01132EB |
25 g |
£12.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
1122-91-4 | |
185.02 | |
p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd | |
4-bromobenzaldehyde |
C7H5BrO | |
ZRYZBQLXDKPBDU-UHFFFAOYSA-N | |
70741 | |
C1=CC(=CC=C1C=O)Br |
Specifications
1122-91-4 | |
C7H5BrO | |
p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd | |
C1=CC(=CC=C1C=O)Br | |
185.02 | |
185.02 | |
4-Bromobenzaldehyde |
95% | |
BrC6H4CHO | |
ZRYZBQLXDKPBDU-UHFFFAOYSA-N | |
4-bromobenzaldehyde | |
70741 | |
95% |
Safety and Handling
Warning
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title