Learn More
4-Bromobenzenediazonium tetrafluoroborate, 96%, Thermo Scientific Chemicals
£42.30 - £152.00
Chemical Identifiers
CAS | 673-40-5 |
---|---|
Molecular Formula | C6H4BBrF4N2 |
Molecular Weight (g/mol) | 270.82 |
MDL Number | MFCD00011894 |
InChI Key | FXTCQUCYDJZGGU-UHFFFAOYSA-N |
Synonym | 4-bromobenzenediazonium tetrafluoroborate, acmc-209nyn, 4-bromobenzene-1-diazonium; tetrafluoroboranuide, boron +3 cation; 4-bromobenzenediazonium; tetrafluoride |
PubChem CID | 2734810 |
IUPAC Name | 4-bromobenzenediazonium;tetrafluoroborate |
SMILES | F[B-](F)(F)F.BrC1=CC=C(C=C1)[N+]#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11491687
|
Thermo Scientific Alfa Aesar
B25670.06 |
5 g |
£42.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401697
|
Thermo Scientific Alfa Aesar
B25670.14 |
25 g |
£152.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.
Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture.
Chemical Identifiers
673-40-5 | |
270.82 | |
FXTCQUCYDJZGGU-UHFFFAOYSA-N | |
2734810 | |
F[B-](F)(F)F.BrC1=CC=C(C=C1)[N+]#N |
C6H4BBrF4N2 | |
MFCD00011894 | |
4-bromobenzenediazonium tetrafluoroborate, acmc-209nyn, 4-bromobenzene-1-diazonium; tetrafluoroboranuide, boron +3 cation; 4-bromobenzenediazonium; tetrafluoride | |
4-bromobenzenediazonium;tetrafluoroborate |
Specifications
673-40-5 | |
C6H4BBrF4N2 | |
UN1759 | |
4-bromobenzenediazonium tetrafluoroborate, acmc-209nyn, 4-bromobenzene-1-diazonium; tetrafluoroboranuide, boron +3 cation; 4-bromobenzenediazonium; tetrafluoride | |
F[B-](F)(F)F.BrC1=CC=C(C=C1)[N+]#N | |
270.82 | |
270.83 | |
4-Bromobenzenediazonium tetrafluoroborate |
∼135°C (decomposition) | |
MFCD00011894 | |
Light and moisture sensitive | |
FXTCQUCYDJZGGU-UHFFFAOYSA-N | |
4-bromobenzenediazonium;tetrafluoroborate | |
2734810 | |
96% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 211-608-2
missing translation for 'tsca' : No
Recommended Storage : Keep cold
- 4-Bromobenzenediazonium tetrafluoroborate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 4-Bromobenzenediazonium tetrafluoroborate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.