missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzoyl chloride, 98%

Product Code. 10447670 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10447670

Brand: Thermo Scientific Acros 106760100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-75-4 | C7H4BrClO | 219.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-75-4
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD00000683
InChI Key DENKGPBHLYFNGK-UHFFFAOYSA-N
Synonym p-bromobenzoyl chloride, benzoyl chloride, 4-bromo, benzoyl chloride, p-bromo, 4-bromobenzoylchloride, p-bromobenzoyl chloride, p-bromo, 4-bromobenzoic acid chloride, p-brombenzoylchlorid, 4bromobenzoyl chloride, p-brc6h4cocl, 4-bromo-benzoylchloride
PubChem CID 68515
IUPAC Name 4-bromobenzoyl chloride
SMILES ClC(=O)C1=CC=C(Br)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 36.0°C to 41.0°C
Color White to Brown
Boiling Point 246.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrC6H4COCl
Quantity 10 g
Beilstein 09,353
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in ethanol,ethyl ether,benzene,ligroin
Formula Weight 219.46
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 4-Bromobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.