missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals

Product Code. 11410483
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.

Solubility
Insoluble in water.

Notes
Sensitive to light. Store in cool dry place in tightly closed container. Avoid excessive heat and light. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-66-0
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.108
MDL Number MFCD00000100
InChI Key PKJBWOWQJHHAHG-UHFFFAOYSA-N
Synonym 4-bromobiphenyl, p-bromobiphenyl, 4-bromo-1,1'-biphenyl, 4-bromodiphenyl, 1,1'-biphenyl, 4-bromo, 4-biphenyl bromide, p-bromodiphenyl, biphenyl, 4-bromo, 4-bromo-biphenyl, p-phenylbromobenzene
PubChem CID 7101
IUPAC Name 1-bromo-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)Br

Specifications

Melting Point 88°C to 92°C
Boiling Point 310°C
Flash Point 143°C (289°F)
Quantity 50 g
Beilstein 1907453
Solubility Information Insoluble in water.
Formula Weight 233.12
Percent Purity ≥98%
Chemical Name or Material 4-Bromobiphenyl

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.