Learn More
4-Bromobutyronitrile, 97%, Thermo Scientific Chemicals
£26.50 - £106.00
Chemical Identifiers
CAS | 5332-06-9 |
---|---|
Molecular Formula | C4H6BrN |
Molecular Weight (g/mol) | 148 |
MDL Number | MFCD00001971 |
InChI Key | CQPGDDAKTTWVDD-UHFFFAOYSA-N |
Synonym | 4-bromobutyronitrile, butanenitrile, 4-bromo, 1-bromo-3-cyanopropane, 3-bromopropyl cyanide, 3-cyanopropyl bromide, gamma-bromobutyronitrile, butyronitrile, 4-bromo, usaf do-6, 4-bromo butyronitrile, trimethylene bromocyanide |
PubChem CID | 21412 |
IUPAC Name | 4-bromobutanenitrile |
SMILES | C(CC#N)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10477860
|
Thermo Scientific Acros
106800050 |
5 mL | Glass bottle |
£26.50
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10487860
|
Thermo Scientific Acros
106800250 |
25 mL | Glass bottle |
£106.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5332-06-9 | |
148 | |
CQPGDDAKTTWVDD-UHFFFAOYSA-N | |
21412 | |
C(CC#N)CBr |
C4H6BrN | |
MFCD00001971 | |
4-bromobutyronitrile, butanenitrile, 4-bromo, 1-bromo-3-cyanopropane, 3-bromopropyl cyanide, 3-cyanopropyl bromide, gamma-bromobutyronitrile, butyronitrile, 4-bromo, usaf do-6, 4-bromo butyronitrile, trimethylene bromocyanide | |
4-bromobutanenitrile |
Specifications
5332-06-9 | |
100.0 | |
1.4800g/mL | |
103°C | |
96% min. (GC) | |
Br(CH2)3CN | |
MFCD00001971 | |
1.48 | |
Solubility in water: immiscible | |
C(CC#N)CBr | |
148 | |
148 | |
Liquid |
96.0 | |
Colorless to Yellow | |
205.0°C | |
Authentic | |
C4H6BrN | |
1.4770 to 1.4790 | |
02, 283 | |
4-bromobutyronitrile, butanenitrile, 4-bromo, 1-bromo-3-cyanopropane, 3-bromopropyl cyanide, 3-cyanopropyl bromide, gamma-bromobutyronitrile, butyronitrile, 4-bromo, usaf do-6, 4-bromo butyronitrile, trimethylene bromocyanide | |
CQPGDDAKTTWVDD-UHFFFAOYSA-N | |
4-bromobutanenitrile | |
21412 | |
97% | |
4-Bromobutyronitrile, 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 226-236-6
missing translation for 'rtecsNumber' : ET8925000
missing translation for 'tsca' : TSCA
- 4-Bromobutyronitrile
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only