missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyryl chloride, 97%, Thermo Scientific Chemicals

£24.60 - £191.00

Chemical Identifiers

CAS 927-58-2
Molecular Formula C4H6BrClO
Molecular Weight (g/mol) 185.445
MDL Number MFCD00000753
InChI Key LRTRXDSAJLSRTG-UHFFFAOYSA-N
Synonym 4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride
PubChem CID 70225
IUPAC Name 4-bromobutanoyl chloride
SMILES C(CC(=O)Cl)CBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11470013
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12094.06
5 g
£24.60
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11480013
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12094.14
25 g
£55.20
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11490013
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12094.22
100 g
£191.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Bromobutyryl chloride is used as a precursor for the synthesis of carolic acid by reacting with ethoxymagnesiomalonic ester. It is widely utilized as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobutyryl chloride is used as a precursor for the synthesis of carolic acid by reacting with ethoxymagnesiomalonic ester. It is widely utilized as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Remove all sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

927-58-2
185.445
LRTRXDSAJLSRTG-UHFFFAOYSA-N
70225
C(CC(=O)Cl)CBr
C4H6BrClO
MFCD00000753
4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride
4-bromobutanoyl chloride

Specifications

927-58-2
102°C to 103°C (40 mmHg)
C4H6BrClO
1.492
UN3265
Moisture sensitive
Miscible with organic solvents. Immiscible with water.
C(CC(=O)Cl)CBr
185.445
185.45
4-Bromobutyryl chloride
1.6
91°C (195°F)
Br(CH2)3COCl
MFCD00000753
1743026
4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride
LRTRXDSAJLSRTG-UHFFFAOYSA-N
4-bromobutanoyl chloride
70225
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 213-154-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only