missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromocinnamic acid, predominantly trans, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.
Solubility
Soluble in alcohol, ethyl acetate, DMSO. Insoluble in water.
Notes
Store at -20°C. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.
Solubility
Soluble in alcohol, ethyl acetate, DMSO. Insoluble in water.
Notes
Store at -20°C. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
Specifications
Melting Point | 260°C to 264°C |
Odor | Odorless |
Quantity | 5 g |
Beilstein | 3538823 |
Solubility Information | Soluble in alcohol,ethyl acetate,DMSO. Insoluble in water. |
Formula Weight | 227.06 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Bromocinnamic acid, predominantly trans |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'