missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoheptane, 97%, Thermo Scientific Chemicals

£46.30 - £148.00

Chemical Identifiers

CAS 998-93-6
Molecular Formula C7H15Br
Molecular Weight (g/mol) 179.10
MDL Number MFCD00039182
InChI Key BNUTXEKPXPZAIT-UHFFFAOYSA-N
Synonym heptane, 4-bromo, 4-bromo-heptane, acmc-20ammr, 4-bromoheptane 5g
PubChem CID 70460
IUPAC Name 4-bromoheptane
SMILES CCCC(Br)CCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381717
View Documents
Thermo Scientific Alfa Aesar
L04807.06
5 g
£46.30
5g
Please to purchase this item. Need a web account? Register with us today!
11391717
View Documents
Thermo Scientific Alfa Aesar
L04807.14
25 g
£148.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Bromoheptane is used in the preparation of 4-nitro-heptane by reacting with sodium nitrite as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoheptane is used in the preparation of 4-nitro-heptane by reacting with sodium nitrite as a reagent.

Solubility
Miscible with carbon tetrachloride, chloroform and benzene. Immiscible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

998-93-6
179.10
BNUTXEKPXPZAIT-UHFFFAOYSA-N
70460
CCCC(Br)CCC
C7H15Br
MFCD00039182
heptane, 4-bromo, 4-bromo-heptane, acmc-20ammr, 4-bromoheptane 5g
4-bromoheptane

Specifications

998-93-6
60°C to 62°C (18 mmHg)
C7H15Br
MFCD00039182
1697144
Miscible with carbon tetrachloride,chloroform and benzene. Immiscible with water.
CCCC(Br)CCC
179.10
179.11
4-Bromoheptane
1.14
40°C (104°F)
1.45
UN1993
heptane, 4-bromo, 4-bromo-heptane, acmc-20ammr, 4-bromoheptane 5g
BNUTXEKPXPZAIT-UHFFFAOYSA-N
4-bromoheptane
70460
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 213-653-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only