Learn More
4-Bromoimidazole, 98%, Thermo Scientific Chemicals
£39.10 - £126.00
Chemical Identifiers
CAS | 2302-25-2 |
---|---|
Molecular Formula | C3H3BrN2 |
Molecular Weight (g/mol) | 146.98 |
MDL Number | MFCD00047021 |
InChI Key | FHZALEJIENDROK-UHFFFAOYSA-N |
Synonym | 4-bromo-1h-imidazole, 4-bromoimidazole, 1h-imidazole, 4-bromo, 4 or 5-bromoimidazole, imidazole, 4-bromo, 4-bromo-3h-imidazole, imidazole, 4 or 5-bromo-6ci,7ci, 5-bromoimidazole, 3kqm, 4-bromo-imidazole |
PubChem CID | 96125 |
IUPAC Name | 5-bromo-1H-imidazole |
SMILES | BrC1=CN=CN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442409
|
Thermo Scientific Alfa Aesar
H64972.06 |
5 g |
£39.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452409
|
Thermo Scientific Alfa Aesar
H64972.14 |
25 g |
£126.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
2302-25-2 | |
146.98 | |
FHZALEJIENDROK-UHFFFAOYSA-N | |
96125 | |
BrC1=CN=CN1 |
C3H3BrN2 | |
MFCD00047021 | |
4-bromo-1h-imidazole, 4-bromoimidazole, 1h-imidazole, 4-bromo, 4 or 5-bromoimidazole, imidazole, 4-bromo, 4-bromo-3h-imidazole, imidazole, 4 or 5-bromo-6ci,7ci, 5-bromoimidazole, 3kqm, 4-bromo-imidazole | |
5-bromo-1H-imidazole |
Specifications
2302-25-2 | |
C3H3BrN2 | |
UN2811 | |
4-bromo-1h-imidazole, 4-bromoimidazole, 1h-imidazole, 4-bromo, 4 or 5-bromoimidazole, imidazole, 4-bromo, 4-bromo-3h-imidazole, imidazole, 4 or 5-bromo-6ci,7ci, 5-bromoimidazole, 3kqm, 4-bromo-imidazole | |
FHZALEJIENDROK-UHFFFAOYSA-N | |
5-bromo-1H-imidazole | |
96125 | |
98% | |
4-Bromoimidazole |
131°C to 135°C | |
MFCD00047021 | |
1615 | |
Slightly soluble in water. | |
BrC1=CN=CN1 | |
146.98 | |
146.97 | |
Solid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : NI3517450
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only