Learn More
4-(Bromomethyl)benzoic acid, 97%, Thermo Scientific Chemicals
£30.90 - £314.00
Chemical Identifiers
CAS | 6232-88-8 |
---|---|
Molecular Formula | C8H7BrO2 |
Molecular Weight (g/mol) | 215.046 |
MDL Number | MFCD00002567 |
InChI Key | CQQSQBRPAJSTFB-UHFFFAOYSA-N |
Synonym | 4-bromomethyl benzoic acid, alpha-bromo-p-toluic acid, benzoic acid, 4-bromomethyl, p-bromomethyl benzoic acid, 4-carboxybenzyl bromide, 4-bromomethyl-benzoic acid, a-bromo-p-toluic acid, p-carboxybenzyl bromide, p-bromomethylbenzoic acid |
PubChem CID | 22599 |
IUPAC Name | 4-(bromomethyl)benzoic acid |
SMILES | C1=CC(=CC=C1CBr)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392116
|
Thermo Scientific Alfa Aesar
L03371.06 |
5 g |
£30.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302126
|
Thermo Scientific Alfa Aesar
L03371.14 |
25 g |
£101.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312126
|
Thermo Scientific Alfa Aesar
L03371.22 |
100 g |
£314.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-(Bromomethyl)benzoic acid acts as an intermediate in the synthesis of eprosartan, which is used as an antihypertensive agent. Further, it is used in the preparation of 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (temoporfin), which is a second generation photosensitizer. In addition to this, it is used in the preparation of 4-(5-arylidene-2,4-dioxothiazolidin-3-yl) methylbenzoic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Bromomethyl)benzoic acid acts as an intermediate in the synthesis of eprosartan, which is used as an antihypertensive agent. Further, it is used in the preparation of 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (temoporfin), which is a second generation photosensitizer. In addition to this, it is used in the preparation of 4-(5-arylidene-2,4-dioxothiazolidin-3-yl) methylbenzoic acids.
Solubility
Soluble in water.
Notes
Incompatible with bases, amines and oxidizing agents.
Chemical Identifiers
6232-88-8 | |
215.046 | |
CQQSQBRPAJSTFB-UHFFFAOYSA-N | |
22599 | |
C1=CC(=CC=C1CBr)C(=O)O |
C8H7BrO2 | |
MFCD00002567 | |
4-bromomethyl benzoic acid, alpha-bromo-p-toluic acid, benzoic acid, 4-bromomethyl, p-bromomethyl benzoic acid, 4-carboxybenzyl bromide, 4-bromomethyl-benzoic acid, a-bromo-p-toluic acid, p-carboxybenzyl bromide, p-bromomethylbenzoic acid | |
4-(bromomethyl)benzoic acid |
Specifications
6232-88-8 | |
C8H7BrO2 | |
UN3261 | |
4-bromomethyl benzoic acid, alpha-bromo-p-toluic acid, benzoic acid, 4-bromomethyl, p-bromomethyl benzoic acid, 4-carboxybenzyl bromide, 4-bromomethyl-benzoic acid, a-bromo-p-toluic acid, p-carboxybenzyl bromide, p-bromomethylbenzoic acid | |
CQQSQBRPAJSTFB-UHFFFAOYSA-N | |
4-(bromomethyl)benzoic acid | |
22599 | |
97% |
223°C to 227°C | |
MFCD00002567 | |
1862870 | |
Soluble in water. | |
C1=CC(=CC=C1CBr)C(=O)O | |
215.046 | |
215.05 | |
4-(Bromomethyl)benzoic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 228-343-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only