missing translation for 'onlineSavingsMsg'
Learn More

[4-(Bromomethyl)benzyl]triphenylphosphonium bromide, 98%, Thermo Scientific™

Product Code. 11408197
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14366-74-6
Molecular Formula C26H23Br2P
Molecular Weight (g/mol) 526.25
MDL Number MFCD00031635
InChI Key KUXBEOGHKWNPTG-UHFFFAOYSA-M
Synonym 4-bromomethyl benzyl triphenylphosphonium bromide, p-bromomethyl benzyltriphenylphosphonium bromide, phosphonium, p-bromomethylbenzyl triphenyl-, bromide, acmc-1c314, wln: e1r d1pr&r&r &e, 4-bromomethyl benzyl triphenylphosphoniumbromide, 4-bromomethyl phenyl methyl-triphenylphosphanium bromide, 4-bromomethyl phenyl methyl triphenylphosphanium bromide, phosphonium, 4-bromomethyl phenyl methyl triphenyl-,bromide 1:1
PubChem CID 197927
IUPAC Name [4-(bromomethyl)phenyl]methyl-triphenylphosphanium;bromide
SMILES [Br-].BrCC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 246°C to 248°C (decomposition)
Quantity 100 g
Beilstein 3820074
Formula Weight 526.25
Percent Purity 98%
Chemical Name or Material [4-(Bromomethyl)benzyl]triphenylphosphonium bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'