missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)phenylacetic acid, 97%, Thermo Scientific Chemicals

Product Code. 10434895
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13737-36-5
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00010632
InChI Key WCOCCXZFEJGHTC-UHFFFAOYSA-N
Synonym 4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid
PubChem CID 4519056
IUPAC Name 2-[4-(bromomethyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 180°C to 182°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula BrCH2C6H4CH2COOH
Quantity 5 g
Formula Weight 229.07
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Bromomethyl)phenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Bromomethyl)phenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'