missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethylbenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 10116673
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66176-39-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.55
MDL Number MFCD00156129
InChI Key QXTQWYZHHMQSQH-UHFFFAOYSA-N
Synonym 4-bromomethyl benzenesulfonyl chloride, 4-bromomethyl benzene-1-sulfonyl chloride, 4-bromomethyl benzenesulfonylchloride, alpha-bromo-p-toluenesulphonyl chloride, alpha-bromo-p-toluenesulfonyl chloride, 4-bromomethyl benzenesulphonyl chloride, 4-bromomethyl benzene-1-sulfonylchloride, buttpark 94\04-08, 4-bromomethyl phenyl chlorosulfone, 4-bromomethylene benzenesulfonyl chloride
PubChem CID 2734409
IUPAC Name 4-(bromomethyl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1CBr)S(=O)(=O)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 69.0°C to 76.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2C6H4SO2Cl
Quantity 25 g
Formula Weight 269.55
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromomethylbenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromomethylbenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'