missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenol, 97%, Thermo Scientific Chemicals

Product Code. 10531911
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-41-2
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.01
MDL Number MFCD00002313
InChI Key GZFGOTFRPZRKDS-UHFFFAOYSA-N
Synonym p-bromophenol, phenol, 4-bromo, p-bromohydroxybenzene, 4-bromo-phenol, p-bromophenic acid, phenol, p-bromo, para-bromophenol, 4-bromo phenol, unii-lao4j0183i, ccris 632
PubChem CID 7808
IUPAC Name 4-bromophenol
SMILES C1=CC(=CC=C1O)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 64°C to 68°C
Color Brown-Pink
Boiling Point 235°C to 236°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OH
Quantity 5 g
Beilstein 06, 198
Merck Index 15, 1439
Solubility Information Solubility in water: 25g/L (25°C). Other solubilities: freely soluble in glacial acetic acid,freely soluble in alcohol,chloroform and ether
Formula Weight 173.01
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'