missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenoxyacetonitrile, 97%, Thermo Scientific™

Product Code. 11480080
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39489-67-3
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.05
MDL Number MFCD00464958
InChI Key PMDDXFGYBKIPKR-UHFFFAOYSA-N
Synonym 2-4-bromophenoxy acetonitrile, 4-bromophenoxyacetonitrile, acmc-1afvu, maybridge3_005625, p-bromo-phenoxyacetonitrile, 4-bromophenoxy-acetonitrile, 4-bromo-phenoxy-acetonitrile, 4-bromophenoxy acetonitrile #, 2-4-bromophenoxy ethanenitrile, 4-bromophenyl oxy acetonitrile
PubChem CID 598823
IUPAC Name 2-(4-bromophenoxy)acetonitrile
SMILES BrC1=CC=C(OCC#N)C=C1

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 212.05
Percent Purity 97%
Chemical Name or Material 4-Bromophenoxyacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'