missing translation for 'onlineSavingsMsg'
Learn More
4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals Product Code.: 10338500

4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals

Product Code. 10338500
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10338500

Brand: Thermo Scientific Acros 221130100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2493-02-9
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00002022
InChI Key CZQIJQFTRGDODI-UHFFFAOYSA-N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 39.0°C to 42.0°C
Color White to Beige
Boiling Point 158.0°C (14.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4NCO
Quantity 10 g
Beilstein 12, 647
Merck Index 15, 1441
Synonym 4-bromophenyl isocyanate, 4-bromophenylisocyanate, p-bromophenyl isocyanate, benzene, 1-bromo-4-isocyanato, p-bromophenylisocyanate, p-bromocarbanil, p-bromophenyl carbonimide, isocyanic acid 4-bromophenyl ester, unii-mgr77m31zw, mgr77m31zw
Solubility Information Solubility in water: .. Other solubilities: soluble in ether
SMILES C1=CC(=CC=C1N=C=O)Br
IUPAC Name 1-bromo-4-isocyanatobenzene
PubChem CID 75609
Formula Weight 198.02
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromophenyl isocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0412657 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number221130500,221130100
Lot NumberA0437722 Certificate TypeCertificate of Analysis Date05/01/2022 Catalog Number221130500,221130100
compliance-icons compliance-icons
Product Identifier
  • 4-Bromophenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 4-Bromophenyl isocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.