Learn More
4-Bromophenylacetonitrile, 99%, Thermo Scientific Chemicals
£25.30 - £55.20
Chemical Identifiers
CAS | 16532-79-9 |
---|---|
Molecular Formula | C8H6BrN |
Molecular Weight (g/mol) | 196.05 |
MDL Number | MFCD00001916 |
InChI Key | MFHFWRBXPQDZSA-UHFFFAOYSA-N |
Synonym | 4-bromophenylacetonitrile, 4-bromobenzyl cyanide, 2-4-bromophenyl acetonitrile, benzeneacetonitrile, 4-bromo, p-bromophenylacetonitrile, 4-bromobenzylcyanide, 4-bromobenzeneacetonitrile, p-bromobenzyl cyanide, p-bromobenzylcyanide, acetonitrile, p-bromophenyl |
PubChem CID | 27914 |
IUPAC Name | 2-(4-bromophenyl)acetonitrile |
SMILES | BrC1=CC=C(CC#N)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10071770
|
Thermo Scientific Acros
154480250 |
25 g | Glass bottle |
£25.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10307570
|
Thermo Scientific Acros
154481000 |
100 g | Glass bottle |
£55.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16532-79-9 | |
196.05 | |
MFHFWRBXPQDZSA-UHFFFAOYSA-N | |
27914 | |
BrC1=CC=C(CC#N)C=C1 |
C8H6BrN | |
MFCD00001916 | |
4-bromophenylacetonitrile, 4-bromobenzyl cyanide, 2-4-bromophenyl acetonitrile, benzeneacetonitrile, 4-bromo, p-bromophenylacetonitrile, 4-bromobenzylcyanide, 4-bromobenzeneacetonitrile, p-bromobenzyl cyanide, p-bromobenzylcyanide, acetonitrile, p-bromophenyl | |
2-(4-bromophenyl)acetonitrile |
Specifications
16532-79-9 | |
100.0 | |
Brown to Colorless | |
Authentic | |
C8H6BrN | |
MFCD00001916 | |
4-bromophenylacetonitrile, 4-bromobenzyl cyanide, 2-4-bromophenyl acetonitrile, benzeneacetonitrile, 4-bromo, p-bromophenylacetonitrile, 4-bromobenzylcyanide, 4-bromobenzeneacetonitrile, p-bromobenzyl cyanide, p-bromobenzylcyanide, acetonitrile, p-bromophenyl | |
MFHFWRBXPQDZSA-UHFFFAOYSA-N | |
2-(4-bromophenyl)acetonitrile | |
27914 | |
99% | |
4-Bromophenylacetonitrile, 99% |
98.5 | |
47°C to 49°C | |
152°C | |
98.5% min. (GC) | |
BrC6H4CH2CN | |
09, 451 | |
Solubility in water: insoluble | |
BrC1=CC=C(CC#N)C=C1 | |
196.05 | |
196.05 | |
Crystalline Mass |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irr
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 240-602-2
missing translation for 'rtecsNumber' : AL8090000
missing translation for 'tsca' : TSCA
- 4-Bromophenylacetonitrile
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only