missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiazole, 97%, Thermo Scientific Chemicals

£43.80 - £153.00

Chemical Identifiers

CAS 34259-99-9
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.02
MDL Number MFCD06657592
InChI Key VDTIGYKLTROQAH-UHFFFAOYSA-N
Synonym 4-bromothiazole, 4-bromo-thiazole, 4-bromo thiazole, thiazole, 4-bromo, 4-bromothiazol, zlchem 498, pubchem4040, ksc222g1h
PubChem CID 2763218
IUPAC Name 4-bromo-1,3-thiazole
SMILES BrC1=CSC=N1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10574038
View Documents
Thermo Scientific Acros
429450010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10155717
View Documents
Thermo Scientific Acros
429450050
5 g Glass bottle
£153.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

34259-99-9
164.02
VDTIGYKLTROQAH-UHFFFAOYSA-N
2763218
BrC1=CSC=N1
C3H2BrNS
MFCD06657592
4-bromothiazole, 4-bromo-thiazole, 4-bromo thiazole, thiazole, 4-bromo, 4-bromothiazol, zlchem 498, pubchem4040, ksc222g1h
4-bromo-1,3-thiazole

Specifications

34259-99-9
100.0
102°C
96% min. (GC)
MFCD06657592
VDTIGYKLTROQAH-UHFFFAOYSA-N
4-bromo-1,3-thiazole
2763218
97%
4-Bromothiazole
96.0
Colorless to Pink
Authentic
C3H2BrNS
4-bromothiazole, 4-bromo-thiazole, 4-bromo thiazole, thiazole, 4-bromo, 4-bromothiazol, zlchem 498, pubchem4040, ksc222g1h
BrC1=CSC=N1
164.02
164.02
Solution
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

Product Identifier
  • 4-Bromothiazole

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only