missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,5-dimethylbenzenesulfonamide, 97%, Thermo Scientific™

Product Code. 11460267
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 219689-73-3
Molecular Formula C8H10ClNO2S
Molecular Weight (g/mol) 219.68
MDL Number MFCD00275263
InChI Key YSDCIOQWHXYICL-UHFFFAOYSA-N
Synonym 4-chloro-2,5-dimethylbenzene-1-sulfonamide, acmc-1chyl, 5-chloro-2-sulphamoyl-p-xylene, 4-chloro-2,5-dimethylbenzenesulphonamide, benzenesulfonamide,4-chloro-2,5-dimethyl, 4-chloranyl-2,5-dimethyl-benzenesulfonamide
PubChem CID 2735730
IUPAC Name 4-chloro-2,5-dimethylbenzenesulfonamide
SMILES CC1=CC(=C(C)C=C1Cl)S(N)(=O)=O

Specifications

Melting Point 190°C to 192°C
Quantity 5 g
Formula Weight 219.69
Percent Purity 97%
Chemical Name or Material 4-Chloro-2,5-dimethylbenzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'