Learn More
4-Chloro-2,6-dimethylphenol, 98%, Thermo Scientific Chemicals
£43.11 - £455.00
Chemical Identifiers
CAS | 1123-63-3 |
---|---|
Molecular Formula | C8H9ClO |
Molecular Weight (g/mol) | 156.609 |
MDL Number | MFCD00020138 |
InChI Key | VWYKSJIPZHRLNO-UHFFFAOYSA-N |
PubChem CID | 70747 |
IUPAC Name | 4-chloro-2,6-dimethylphenol |
SMILES | CC1=CC(=CC(=C1O)C)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11440234
|
Thermo Scientific Alfa Aesar
A13649.09 |
10 g |
£43.11
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450234
|
Thermo Scientific Alfa Aesar
A13649.18 |
50 g |
£160.80
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460234
|
Thermo Scientific Alfa Aesar
A13649.30 |
250 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1123-63-3 | |
156.609 | |
VWYKSJIPZHRLNO-UHFFFAOYSA-N | |
4-chloro-2,6-dimethylphenol |
C8H9ClO | |
MFCD00020138 | |
70747 | |
CC1=CC(=CC(=C1O)C)Cl |
Specifications
1123-63-3 | |
C8H9ClO | |
1908282 | |
VWYKSJIPZHRLNO-UHFFFAOYSA-N | |
4-chloro-2,6-dimethylphenol | |
70747 | |
98% |
80°C to 83°C | |
MFCD00020138 | |
4-chloro-2,6-xylenol, phenol, 4-chloro-2,6-dimethyl, 2,6-dimethyl-4-chlorophenol, 4-chloro-2,6-dimethyl-phenol, 2,6-xylenol, 4-chloro, pubchem2657, acmc-1c4bc, phenol,4-chloro-2,6-dimethyl, 4-chloro-2,6-xylenol oh=1, 4-chloro-2,6-dimethylphenol | |
CC1=CC(=CC(=C1O)C)Cl | |
156.609 | |
156.61 | |
4-Chloro-2,6-dimethylphenol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 214-376-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
- 4-Chloro-2,6-dimethylphenol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 4-Chloro-2,6-xylenol
RUO – Research Use Only