missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoro-5-nitrobenzoic acid, 98%, Thermo Scientific™

Product Code. 15440619
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35112-05-1
Molecular Formula C7H3ClFNO4
Molecular Weight (g/mol) 219.552
MDL Number MFCD04115725
InChI Key KAONECPATVTFST-UHFFFAOYSA-N
Synonym 4-chloro-2-fluoro-5-nitrobenzoic acid, 4-chloro-2-fluoro-5-nitrobenzoicacid, 4-chloro-2-fluoro-5-nitro-benzoic acid, 2-fluoro-4-chloro-5-nitrobenzoic acid, benzoic acid, 4-chloro-2-fluoro-5-nitro, pubchem22286, acmc-1ah9o, ksc573m6p, 5-carboxy-2-chloro-4-fluoronitrobenzene
PubChem CID 3774553
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])Cl)F)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 219.55
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluoro-5-nitrobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'