missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-fluorobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Chloro-2-fluorobenzylamine hydrochloride It is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloro-2-fluorobenzylamine hydrochloride It is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.
Solubility
It is soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
4-Chloro-2-fluorobenzylamine hydrochloride It is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.
Solubility
It is soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Specifications
Melting Point | 243°C to 245°C |
Quantity | 1 g |
Solubility Information | It is soluble in water. |
Formula Weight | 196.05 |
Percent Purity | 97% |
Chemical Name or Material | 4-Chloro-2-fluorobenzylamine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'