missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-isopropyl-5-methylphenol, 99%, Thermo Scientific Chemicals

£38.19 - £105.53

Chemical Identifiers

CAS 89-68-9
Molecular Formula C10H13ClO
Molecular Weight (g/mol) 184.66
MDL Number MFCD00002326
InChI Key KFZXVMNBUMVKLN-UHFFFAOYSA-N
Synonym chlorothymol, 6-chlorothymol, 4-chloro-2-isopropyl-5-methylphenol, chlorthymol, 4-chlorothymol, thymol, 6-chloro, phenol, 4-chloro-5-methyl-2-1-methylethyl, caswell no. 216, 4-chloro-6-isopropyl-3-methylphenol, unii-lj25ti0cvt
PubChem CID 6982
IUPAC Name 4-chloro-5-methyl-2-propan-2-ylphenol
SMILES CC(C)C1=CC(Cl)=C(C)C=C1O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11404987
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21906.14
25 g
£38.19
25g
Please to purchase this item. Need a web account? Register with us today!
11414987
View Documents
Thermo Scientific Alfa Aesar
B21906.22
100 g
£105.53
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

89-68-9
184.66
KFZXVMNBUMVKLN-UHFFFAOYSA-N
6982
CC(C)C1=CC(Cl)=C(C)C=C1O
C10H13ClO
MFCD00002326
chlorothymol, 6-chlorothymol, 4-chloro-2-isopropyl-5-methylphenol, chlorthymol, 4-chlorothymol, thymol, 6-chloro, phenol, 4-chloro-5-methyl-2-1-methylethyl, caswell no. 216, 4-chloro-6-isopropyl-3-methylphenol, unii-lj25ti0cvt
4-chloro-5-methyl-2-propan-2-ylphenol

Specifications

89-68-9
259°C to 263°C
MFCD00002326
Light sensitive
chlorothymol, 6-chlorothymol, 4-chloro-2-isopropyl-5-methylphenol, chlorthymol, 4-chlorothymol, thymol, 6-chloro, phenol, 4-chloro-5-methyl-2-1-methylethyl, caswell no. 216, 4-chloro-6-isopropyl-3-methylphenol, unii-lj25ti0cvt
CC(C)C1=CC(Cl)=C(C)C=C1O
184.66
184.67
4-Chloro-2-isopropyl-5-methylphenol
59°C to 61°C
C10H13ClO
2084453
14,2170
KFZXVMNBUMVKLN-UHFFFAOYSA-N
4-chloro-5-methyl-2-propan-2-ylphenol
6982
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 201-930-1

missing translation for 'rtecsNumber' : XP2620000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only