Learn More
4-Chloro-2-methoxyphenol, 97%, Thermo Scientific Chemicals
£31.20 - £87.10
Chemical Identifiers
CAS | 16766-30-6 |
---|---|
Molecular Formula | C7H7ClO2 |
Molecular Weight (g/mol) | 158.58 |
MDL Number | MFCD00070774 |
InChI Key | FVZQMMMRFNURSH-UHFFFAOYSA-N |
PubChem CID | 28050 |
IUPAC Name | 4-chloro-2-methoxyphenol |
SMILES | COC1=CC(Cl)=CC=C1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11432557
|
Thermo Scientific Alfa Aesar
A14806.06 |
5 g |
£31.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442557
|
Thermo Scientific Alfa Aesar
A14806.14 |
25 g | |||||||
Description
4-Chloro-2-methoxyphenol, is used as an intermediate in pharmaceutical research, pharmaceutical chemicals and reagents. It is also used as an application intermediate. it can be used to produce 4-Chloro-2-methoxy-6-nitrophenol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chloro-2-methoxyphenol, is used as an intermediate in pharmaceutical research, pharmaceutical chemicals and reagents. It is also used as an application intermediate. it can be used to produce 4-Chloro-2-methoxy-6-nitrophenol.
Solubility
It is slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases.
Chemical Identifiers
16766-30-6 | |
158.58 | |
FVZQMMMRFNURSH-UHFFFAOYSA-N | |
4-chloro-2-methoxyphenol |
C7H7ClO2 | |
MFCD00070774 | |
28050 | |
COC1=CC(Cl)=CC=C1O |
Specifications
16766-30-6 | |
1.304 | |
>110°C (230°F) | |
C7H7ClO2 | |
MFCD00070774 | |
4-chloroguaiacol, phenol, 4-chloro-2-methoxy, 4-chloro-2-methoxy-phenol, 2-methoxy-4-chloro-phenol, pubchem3665, acmc-209dwx, #, phenol,4-chloro-2-methoxy, 4-chloranyl-2-methoxy-phenol, fvzqmmmrfnursh-uhfffaoysa | |
FVZQMMMRFNURSH-UHFFFAOYSA-N | |
4-chloro-2-methoxyphenol | |
28050 | |
97% |
16°C to 17°C | |
241°C | |
Phenol-like | |
1.5595 | |
1940516 | |
It is slightly soluble in water. | |
COC1=CC(Cl)=CC=C1O | |
158.58 | |
158.59 | |
4-Chloro-2-methoxyphenol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only