missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylbenzyl alcohol, 97%, Thermo Scientific™

Product Code. 11461850
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129716-11-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD06201140
InChI Key AHXBDGJNZJKLNE-UHFFFAOYSA-N
Synonym 4-chloro-2-methylphenyl methanol, 4-chloro-2-methylbenzyl alcohol, benzenemethanol, 4-chloro-2-methyl, acmc-20aaxd, 4-chloro-2-methylbenzylalcohol, 4-chloro-2-methyl-phenyl-methanol
PubChem CID 2757669
IUPAC Name (4-chloro-2-methylphenyl)methanol
SMILES CC1=C(C=CC(=C1)Cl)CO

Specifications

Melting Point 60°C to 64°C
Quantity 1 g
Formula Weight 156.61
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylbenzyl alcohol
compliance-icons
Product Identifier
  • 4-Chloro-2-methylbenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Chloro-2-methylbenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'