missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylquinoline, 97%, Thermo Scientific Chemicals

Product Code. 11487227
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4295-06-1
MDL Number MFCD00006757

Specifications

Melting Point ∼25°C
Density 0.881
Boiling Point 269°C to 270°C
Flash Point >110°C (230°F)
Molecular Formula C10H8ClN
Refractive Index 1.622
Quantity 25 g
Beilstein 3522
Synonym 4-chloroquinaldine, quinoline, 4-chloro-2-methyl, 4-chloro-2-methyl-quinoline, 2-methyl-4-chloroquinoline, quinaldine, 4-chloro, 4-chloro-quinaldine, acmc-20a2ui, 4-chloroquinaldine hcl, quinoline,4-chloro-2-methyl
Solubility Information Slightly soluble in water.
InChI Key HQAIROMRVBVWSK-UHFFFAOYSA-N
SMILES CC1=NC2=CC=CC=C2C(=C1)Cl
IUPAC Name 4-chloro-2-methylquinoline
Molecular Weight (g/mol) 177.63
PubChem CID 77973
Formula Weight 177.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylquinoline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'