Learn More
4-Chloro-3,5-dimethylphenol, 98+%, Thermo Scientific Chemicals
£17.80 - £49.70
Chemical Identifiers
CAS | 88-04-0 |
---|---|
Molecular Formula | C8H9ClO |
Molecular Weight (g/mol) | 156.609 |
MDL Number | MFCD00002324 |
InChI Key | OSDLLIBGSJNGJE-UHFFFAOYSA-N |
Synonym | chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson |
PubChem CID | 2723 |
ChEBI | CHEBI:34393 |
IUPAC Name | 4-chloro-3,5-dimethylphenol |
SMILES | CC1=CC(=CC(=C1Cl)C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11425077
|
Thermo Scientific Alfa Aesar
B21964.22 |
100 g |
£17.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11435077
|
Thermo Scientific Alfa Aesar
B21964.36 |
500 g |
£49.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in hospitals and households for disinfection and sanitation. It is used in wound-cleansing applications and household antiseptics such as Dettol liquid, cream and ointments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in hospitals and households for disinfection and sanitation. It is used in wound-cleansing applications and household antiseptics such as Dettol liquid, cream and ointments.
Solubility
Soluble in benzene, toluene, glycerin, chloroform, acetone, isopropanol, alcohol, ether, terpenes and in solutions of alkali hydroxides. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, strong bases.
Chemical Identifiers
88-04-0 | |
156.609 | |
OSDLLIBGSJNGJE-UHFFFAOYSA-N | |
2723 | |
4-chloro-3,5-dimethylphenol |
C8H9ClO | |
MFCD00002324 | |
chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson | |
CHEBI:34393 | |
CC1=CC(=CC(=C1Cl)C)O |
Specifications
88-04-0 | |
∼246°C | |
Odorless | |
MFCD00002324 | |
14,2176 | |
Soluble in benzene,toluene,glycerin,chloroform,acetone,isopropanol,alcohol,ether,terpenes and in solutions of alkali hydroxides. Slightly soluble in water. | |
CC1=CC(=CC(=C1Cl)C)O | |
156.609 | |
CHEBI:34393 | |
≥98% |
112°C to 116°C | |
138°C (280°F) | |
C8H9ClO | |
1862539 | |
chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson | |
OSDLLIBGSJNGJE-UHFFFAOYSA-N | |
4-chloro-3,5-dimethylphenol | |
2723 | |
156.61 | |
4-Chloro-3,5-dimethylphenol |
Safety and Handling
GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
H302-H315-H317-H319
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 201-793-8
missing translation for 'rtecsNumber' : ZE6850000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only