missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-cyanobenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15489767
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871332-95-5
Molecular Formula C7H5BClNO2
Molecular Weight (g/mol) 181.38
MDL Number MFCD08056360
InChI Key PRPLADNLEYNYFZ-UHFFFAOYSA-N
Synonym 4-chloro-3-cyanophenyl boronic acid, 5-borono-2-chlorobenzonitrile, 4-chloro-3-cyanobenzeneboronic acid, 4-chloro-3-cyano-phenyl boronic acid, 4-chloro-3-cyanophenylboronicacid, pubchem17104, acmc-209qho, ksc447q0n, 4-chloro-3-cyanobenzeneboronic acid 97+%
PubChem CID 44119831
IUPAC Name (4-chloro-3-cyanophenyl)boronic acid
SMILES OB(O)C1=CC(C#N)=C(Cl)C=C1

Specifications

Melting Point 300°C to 304°C
Quantity 1 g
Formula Weight 181.39
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-cyanobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'