Learn More
4-Chloro-3-ethoxy-2-fluorobenzoyl chloride, 97%, Thermo Scientific™
£26.92 - £96.40
Chemical Identifiers
CAS | 1323966-42-2 |
---|---|
Molecular Formula | C9H7Cl2FO2 |
Molecular Weight (g/mol) | 237.051 |
MDL Number | MFCD19687216 |
InChI Key | XTWYKHZMPKQVSM-UHFFFAOYSA-N |
PubChem CID | 66523586 |
IUPAC Name | 4-chloro-3-ethoxy-2-fluorobenzoyl chloride |
SMILES | CCOC1=C(C=CC(=C1F)C(=O)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15428365
|
Thermo Scientific Alfa Aesar
H33199.MD |
250 mg | |||||||
15475987
|
Thermo Scientific Alfa Aesar
H33199.03 |
1 g |
£96.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1323966-42-2 | |
237.051 | |
XTWYKHZMPKQVSM-UHFFFAOYSA-N | |
4-chloro-3-ethoxy-2-fluorobenzoyl chloride |
C9H7Cl2FO2 | |
MFCD19687216 | |
66523586 | |
CCOC1=C(C=CC(=C1F)C(=O)Cl)Cl |
Specifications
1323966-42-2 | |
MFCD19687216 | |
Moisture sensitive | |
CCOC1=C(C=CC(=C1F)C(=O)Cl)Cl | |
237.051 | |
237.06 | |
4-Chloro-3-ethoxy-2-fluorobenzoyl chloride |
C9H7Cl2FO2 | |
UN3261 | |
XTWYKHZMPKQVSM-UHFFFAOYSA-N | |
4-chloro-3-ethoxy-2-fluorobenzoyl chloride | |
66523586 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures