missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methoxyaniline, 98%, Thermo Scientific™

£19.00 - £205.67

Chemical Identifiers

CAS 13726-14-2
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.60
MDL Number MFCD00672967
InChI Key LNKBDFVSILQKSI-UHFFFAOYSA-N
Synonym 3-methoxy-4-chloroaniline, 4-chloro-3-methoxyphenylamine, 4-chloro-m-anisidine, 2-chloro-5-aminoanisole, 5-amino-2-chloroanisole, 4-chloro-3-methoxy aniline, 4-chloro-3-methoxy-phenylamine, benzenamine, 4-chloro-3-methoxy, pubchem3659, acmc-1chua
PubChem CID 13103692
IUPAC Name 4-chloro-3-methoxyaniline
SMILES COC1=C(Cl)C=CC(N)=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13438628
View Documents Promotion Details
Thermo Scientific Acros
448800010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13448628
View Documents Promotion Details
Thermo Scientific Acros
448800050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13458628
View Documents Promotion Details
Thermo Scientific Acros
448800250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

13726-14-2
157.60
LNKBDFVSILQKSI-UHFFFAOYSA-N
13103692
COC1=C(Cl)C=CC(N)=C1
C7H8ClNO
MFCD00672967
3-methoxy-4-chloroaniline, 4-chloro-3-methoxyphenylamine, 4-chloro-m-anisidine, 2-chloro-5-aminoanisole, 5-amino-2-chloroanisole, 4-chloro-3-methoxy aniline, 4-chloro-3-methoxy-phenylamine, benzenamine, 4-chloro-3-methoxy, pubchem3659, acmc-1chua
4-chloro-3-methoxyaniline

Specifications

13726-14-2
97.5% min. (GC)
C7H8ClNO
MFCD00672967
Solubility in water: insoluble. Other solubilities: soluble in methanol
COC1=C(Cl)C=CC(N)=C1
157.60
157.6
4-Chloro-3-methoxyaniline
78°C to 81°C
Glass bottle
ClC6H3(OCH3)NH2
3-methoxy-4-chloroaniline, 4-chloro-3-methoxyphenylamine, 4-chloro-m-anisidine, 2-chloro-5-aminoanisole, 5-amino-2-chloroanisole, 4-chloro-3-methoxy aniline, 4-chloro-3-methoxy-phenylamine, benzenamine, 4-chloro-3-methoxy, pubchem3659, acmc-1chua
LNKBDFVSILQKSI-UHFFFAOYSA-N
4-chloro-3-methoxyaniline
13103692
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Danger

Product Identifier
  • 4-Chloro-3-methoxyaniline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only