missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylphenol, 99%, Thermo Scientific Chemicals

£20.90 - £43.40

Chemical Identifiers

CAS 59-50-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00002323
InChI Key CFKMVGJGLGKFKI-UHFFFAOYSA-N
Synonym chlorocresol, 4-chloro-m-cresol, baktol, parol, p-chloro-m-cresol, ottafact, phenol, 4-chloro-3-methyl, 4-chloro-3-cresol, candaseptic, baktolan
PubChem CID 1732
ChEBI CHEBI:34395
IUPAC Name 4-chloro-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11395129
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17823.22
100 g
£20.90
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11305139
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17823.36
500 g
£43.40
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

59-50-7
142.582
CFKMVGJGLGKFKI-UHFFFAOYSA-N
1732
4-chloro-3-methylphenol
C7H7ClO
MFCD00002323
chlorocresol, 4-chloro-m-cresol, baktol, parol, p-chloro-m-cresol, ottafact, phenol, 4-chloro-3-methyl, 4-chloro-3-cresol, candaseptic, baktolan
CHEBI:34395
CC1=C(C=CC(=C1)O)Cl

Specifications

59-50-7
234°C to 236°C
C7H7ClO
UN3437
14,2133
CFKMVGJGLGKFKI-UHFFFAOYSA-N
4-chloro-3-methylphenol
1732
142.59
4-Chloro-3-methylphenol
64°C to 68°C
118°C (244°F)
MFCD00002323
1237629
chlorocresol, 4-chloro-m-cresol, baktol, parol, p-chloro-m-cresol, ottafact, phenol, 4-chloro-3-methyl, 4-chloro-3-cresol, candaseptic, baktolan
CC1=C(C=CC(=C1)O)Cl
142.582
CHEBI:34395
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H312-H317
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P310-P301+P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID

missing translation for 'rtecsNumber' : GO7100000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only