missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-3-(trifluoromethoxy)aniline, 97%, Thermo Scientific™ Product Code.: 15414377

4-Chloro-3-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 15414377
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15414377

Brand: Thermo Scientific Alfa Aesar H31752.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Chloro-3-(trifluoromethoxy)aniline is widely utilized as an intermediate in organic synthesis.

Notes
Store in a cool and dry place. Keep the container tightly sealed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97608-50-9
Molecular Formula C7H5ClF3NO
Molecular Weight (g/mol) 211.57
MDL Number MFCD09832255
InChI Key RWTSQATWCKAOBB-UHFFFAOYSA-N
PubChem CID 13405086
IUPAC Name 4-chloro-3-(trifluoromethoxy)aniline
SMILES NC1=CC(OC(F)(F)F)=C(Cl)C=C1

Specifications

Quantity 1 g
Refractive Index 1.494
UN Number UN2810
Synonym 4-chloro-3-trifluoromethoxy aniline, 4-chloro-3-trifluoromethoxy benzenamine, 4-chloro-3-trifluoromethoxy phenylamine, benzenamine,4-chloro-3-trifluoromethoxy, benzenamine, 4-chloro-3-trifluoromethoxy, 1-chloro-4-nitro-2-trifluoromethoxy benzene
Formula Weight 211.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-3-(trifluoromethoxy)aniline, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.