missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine, 97%, Thermo Scientific™

Product Code. 10262392
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 306934-78-1
Molecular Formula C13H9ClN2S
Molecular Weight (g/mol) 260.74
MDL Number MFCD02179828
InChI Key LRTKDTZXCFGPEU-UHFFFAOYSA-N
Synonym 4-chloro-5-methyl-6-phenylthieno 2,3-d pyrimidine, 4-chloro-5-methyl-6-phenylthiopheno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-methyl-6-phenyl, 4-chloranyl-5-methyl-6-phenyl-thieno 2,3-d pyrimidine
PubChem CID 2735772
IUPAC Name 4-chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine
SMILES CC1=C(SC2=NC=NC(Cl)=C12)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 260.75
Percent Purity 97%
Chemical Name or Material 4-chloro-5-methyl-6-phenylthieno[2, 3-d]pyrimidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.