missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-5-methylbenzene-1,2-diamine, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Specifications
CAS Min % | 95.0 |
Quantity | 1 g |
Packaging | Amber glass bottle |
Assay Percent Range | 95% |
Synonym | 1,2-benzenediamine,4-chloro-5-methyl, 4-chloro-5-methyl-1,2-phenylenediamine, 4mrh, pubchem22761, 2-chloro-4,5-diaminotoluene, 2-amino-4-chloro-5-methylphenyl amine, 4-chloro-5-methyl-benzene-1,2-diamine, 5-chloro-4-methyl-1,2-phenylenediamine, 1,2-benzenediamine, 4-chloro-5-methyl, 4-chloranyl-5-methyl-benzene-1,2-diamine |
Formula Weight | 156.62 |
Percent Purity | 95% |
Chemical Name or Material | 4-Chloro-5-methylbenzene-1, 2-diamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Chloro-5-methylbenzene-1,2-diamine, ≥95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction