missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-6,7-dimethoxyquinazoline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
A useful synthetic intermediate in the preparation of epidermal growth factors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A useful synthetic intermediate in the preparation of epidermal growth factors.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
A useful synthetic intermediate in the preparation of epidermal growth factors.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 184°C to 187°C |
Quantity | 5 g |
Solubility Information | Insoluble in water. |
Formula Weight | 224.65 |
Percent Purity | 98% |
Chemical Name or Material | 4-Chloro-6,7-dimethoxyquinazoline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'